Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-APOL6, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP120884

Catalog No. NBP120884


Add to cart

Description

Description

APOL6 Polyclonal antibody specifically detects APOL6 in Mouse samples. It is validated for Peptide ELISA.
Specifications

Specifications

APOL6
0.5 mg/mL
Peptide ELISA Detection limit 1:4000
XP_001481349.1
apolipoprotein L, 6, apolipoprotein L6, Apolipoprotein L-VI, APOLVI, APOL-VI, DKFZp667M075, FLJ38562, FLJ90164, MGC57495
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected from sequence similarity: Mouse.
ELISA
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
APOL6
Peptide with sequence C-KDLKAANPTELAE corresponding to internal region according to Uniprot Mouse XP_001481349.1.
Immunogen affinity purified
RUO
Primary
80830
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only