Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein L6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523622

Catalog No. PIPA523622

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles.
Specifications

Specifications

Apolipoprotein L6
Polyclonal
PBS with 0.09% sodium azide
Q9BWW8
APOL-VI, APOLVI, apolipoprotein L-VI, apolipoprotein L6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80830
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
APOL6
Liquid
APOL6
KLH conjugated synthetic peptide between 126-155 amino acids from the central region of human APOL6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit