Learn More
ApoA1 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127156
Catalog No. PA127156
Add to cart
Description
Description
PA1-27156 detects Apolipoprotein A I from hamster samples. PA1-27156 has been successfully used in Western blot, double immunodiffusion and radial immunodiffusion procedures. The predicted molecular weight is 31 kDa. The PA1-27156 immunogen is full length, native protein purified from hamster plasma high density lipoprotein.
Apolipoprotein A I promotes cholesterol efflux from tissues to the liver for excretion. Apolipoprotein A I is the major protein component of high density lipoprotein (HDL) in the plasma. Synthesized in the liver and small intestine, it consists of two identical chains of 77 amino acids; an 18 amino acid signal peptide is removed co-translationally and a 6 amino acid propeptide is cleaved post-translationally. Apolipoprotein A I is a cofactor for lecithin cholesterolacyltransferase (LCAT) which is responsible for the formation of most plasma cholesteryl esters. Defects in the Apolipoprotein A I gene are associated with HDL deficiency and Tangier disease.
Specifications
Specifications
ApoA1
Polyclonal
antibody with 100U/mL Penicillin and 0.05% sodium azide, 10μg/mL Streptomycin, 25ng/mL Amphotericin B
P02647
Apoa1
Full length, native protein purified from hamster plasma high density lipoprotein.
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100772590, 335
Immunodiffusion, Western Blot
Unconjugated
APOA1
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Hamster, Human