Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoA2 Goat anti-Human, HRP, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA128972

Catalog No. PA128972


Add to cart

Description

Description

PA1-28972 detects Apo AII from human samples. PA1-28972 has been successfully used in Western blot and ELISA applications. Predicted molecular weight: 11 kDa. The PA1-28972 immunogen is: Full length protein (Human Apolipoprotein A II). Store at 4°C short term. For extended storage, aliquot and store at -20°C or below. Avoid freeze-thaw cycles.

Specifications

Specifications

ApoA2
Polyclonal
HRP
APOA2
Liquid
Goat
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
336
ELISA, Western Blot
1 mg/mL
PBS with 10mg/mL BSA and 0.01% thimerosal; pH 7.4
P02652
APOA2
Full length protein (Human Apolipoprotein A II).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit