Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoA2 Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA130491

Catalog No. PA130491


Add to cart

Description

Description

PA1-30491 detects Apo AII from human samples. PA1-30491 has been successfully used in ELISA and Western blot applications. The PA1-30491 immunogen is: Full length native protein (purified) (Human).

This gene encodes apolipoprotein (apo-) A-II, which is the second most abundant protein of the high density lipoprotein particles. The protein is found in plasma as a monomer, homodimer, or heterodimer with apolipoprotein D. Defects in this gene may result in apolipoprotein A-II deficiency or hypercholesterolemia.
Specifications

Specifications

ApoA2
Polyclonal
whole serum with 0.1% sodium azide
P02652
APOA2
Full length native protein (purified) (Human).
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
336
ELISA, Western Blot
Unconjugated
APOA2
Liquid
Sheep
IgG
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit