Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoA2 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA126900

Catalog No. PA126900


Description

Description

PA1-26900 detects Apolipoprotein A II from human samples.

ApoA2 Polyclonal antibody specifically detects ApoA2 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ApoA2
Polyclonal
Unconjugated
APOA2
Alp-2; APO A2; APOA2; Apoa-2; ApoAII; Apo-AII; ApoA-II; Apolipoprotein; Apolipoprotein A2; Apolipoprotein AII; apolipoprotein A-II; Apolipoprotein A-II(1-76); Hdl-1; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II
Goat
Antigen affinity chromatography
RUO
336
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
0.125M sodium borate with 0.075M NaCl, 5mM EDTA and 0.01% sodium azide; pH 8.0
P02652
APOA2
Native human apoLipoprotein type A-II.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.