Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoC3 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA122562

Catalog No. PA122562


Add to cart

Description

Description

PA1-22562 detects apolipoprotein C3 in human samples. PA1-22562 has been successfully used in ELISA, immunohistochemistry (paraffin), immunoprecipitation, and Western blot procedures. Typically there is less than 1% cross-reactivity against other apolipoprotein determined in ELISA The PA1-22562 immunogen is purified full length native apolipoprotein type C-III. Store vial at 4°C for short-term use. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing.

At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL and VLDL particles in human serum are often used to estimate an individuals ft. relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins (especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease than measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.
Specifications

Specifications

ApoC3
Polyclonal
Unconjugated
APOC3
Liquid
APOC3
Purified full length native apolipoprotein type C-III.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
0.125M sodium borate with 5mM EDTA and 0.01% sodium azide; pH 8
P02656
APOCIII, MGC150353
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
345
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit