Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Apolipoprotein M/ApoM, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128460

Catalog No. NBP128460

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Apolipoprotein M/ApoM
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000
NP_061286.1
apolipoprotein M, apo-M, ApoM, G3A, HSPC336, MGC22400, NG20, NG20-like protein, Protein G3a
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat.
Western Blot, ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
APOM
Peptide with sequence C-KGNMELRTEGRPD corresponding to internal region according to Uniprot Mouse NP_061286.1.
Immunogen affinity purified
RUO
Primary
55937
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only