Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APPL1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA15890

Catalog No. PA15890


Add to cart

Description

Description

PA1-5890 detects APPL1 in human and mouse samples. PA1-5890 has been successfully used in immunofluorescence, immunohistochemistry (Frozen) and western blotting. The PA1-5890 immunogen is purified APPL1 fusion protein.

The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signalling and insulin signalling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
Specifications

Specifications

APPL1
Polyclonal
whole serum with 0.1% sodium azide
Q9UKG1, Q8K3H0
DIP13alpha
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
APPL1
Liquid
APPL1
Purified APPL1 fusion protein.
50 μL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
26060, 72993
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit