Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APRT, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522040

Catalog No. PIPA522040

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA5-22040 targets APRT in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples.The PA5-22040 immunogen is recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 84 (P07741) of APRT.
Specifications

Specifications

APRT
Polyclonal
Unconjugated
APRT
Purified
APRT
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 84 (P07741) of APRT
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P07741
AMP, AMP diphosphorylase, AMP pyrophosphorylase, transphosphoribosidase
Rabbit
IgG
100μL
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
353
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit