Learn More
Aquaporin Adipose-Specific, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier:  Thermo Scientific PA128172
Catalog No. PIPA128172
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Aquaporins/major intrinsic protein (MIP) are a family of water-selective membrane channels. Aquaporin 7 has greater sequence similarity with AQP3 and AQP9 and they may be a subfamily. Aquaporin 7 and AQP3 are at the same chromosomal location suggesting that 9p13 may be a site of an aquaporin cluster. Aquaporin 7 facilitates water, glycerol and urea transport. It may play an important role in sperm function.
PA1-28172 detects Aquaporin Adipose Specific from human samples.PA1-28172 has been successfully used in Western blot applications.The PA1-28172 immunogen is: Synthetic peptide, corresponding to 20 N-terminal amino acids (predicted to be cytoplasmic) of Human Aquaporin, Adipose Specific. The peptide is unique to Aquaporin, adipose specific, without significant homology to any other A
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aquaporin Adipose Specific
Polyclonal
AQP7
RP11-311H10.6, AQP7L, AQP9, AQPap, MGC149555, MGC149556
Chicken
25 μL
Primary
Human
Antibody
IgY
ELISA, Western Blot
Unconjugated
O14520
AQP7
Synthetic peptide, corresponding to 20 N-terminal amino acids (predicted to be cytoplasmic) of Human Aquaporin, Adipose Specific. The peptide is unique to Aquaporin, adipose specific, without significant homology to any other Aquaporins.
RUO
364
Store at -20°C, Avoid Freeze/Thaw Cycles
Serum
Recommendations
Recommendations