Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arc Mouse, Unlabeled, Clone: 40, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612603

 View more versions of this product

Catalog No. BDB612603


Add to cart

Description

Description

Immediate early genes (IEGs) are the first to be transcribed upon cellular stimulation. Two groups of these genes have been characterized: those which encode transcription factors that indirectly influence cellular activity, and those which encode effector proteins that directly affect cellular activity. Arc (Activity-Regulated Cytoskeleton-associated protein) encodes an effector protein associated with the actin cytoskeletal matrix. In activated neurons of the hippocampus, Arc gene expression is rapidly induced. Arc protein is concentrated in the actin cortex,
adjacent to the plasma membrane of the cell body and dendrites. Arc has a putative role in the maintenance of neuronal plasticity and in activity-dependent changes in dendrite function. Cocaine and D2 receptor antagonists are known to trigger the production and localization of Arc to dendrites and neuronal cell bodies.

Western Blotting

Specifications

Specifications

Arc
40
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Activity-Regulated Cytoskeleton-associated protein
Affinity Purified
Rat Arc aa. 264-385
150μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.