Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARC Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129149

Catalog No. PA129149

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-29157 detects Caspase 3 from rat, rabbit, human, and mouse samples. PA1-29157 has been successfully used in immunoprecipitation, immunohistochemistry (paraffin tissue), and Western blot applications. Possible positive controls are Jurkat cells, and t

ARC (also known as NOL3 - Nucleolar protein 3) has two isoforms. Isoform 1 is involved in RNA splicing. Isoform 2 functions as an apoptosis repressor that blocks multiple modes of cell death. ARC inhibits extrinsic apoptotic pathways two different ways. The first is by interacting with FAS and FADD upon FAS activation blocking death-inducing signaling complex (DISC) assembly. The second way is by interacting with CASP8 in a mitochondria localization-and phosphorylation-dependent manner, which in turn limits the amount of soluble CASP8 avaliable for DISC-mediated activation. ARC also inhibits intrinsic apoptotic pathways in response to a wide range of stresses through interactions with BAX. This results in BAX inactivation and prevents mitochondrial dysfunction and release of pro-apoptotic factors. Mutations affecting the gene can results in myoclonus, familiam cortical.
Specifications

Specifications

ARC
Polyclonal
Unconjugated
NOL3
Liquid
NOL3
Synthetic peptide corresponding to the C-terminus of Human A
Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
O60936
Apoptosis Repressor with CARD, Nop30, Nucleolar protein of 30 kDa, Nucleolar protein 3, Muscle-enriched cytoplasmic protein, Myp
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
8996
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit