Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amphiregulin, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127954

Catalog No. PIPA127954

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Bifunctional growth-modulating glycoprotein. Inhibits growth of several human carcinoma cells in culture and stimulates proliferation of human fibroblasts and certain other tumor cells. The protein encoded by this gene is a member of the epidermal growth

PA1-27954 detects AREG from human and chimpanzee samples.PA1-27954 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27592 immunogen is a synthetic peptide corresponding to human Amphiregulin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AREG
Polyclonal
AREG
AR, CRDGF, MGC13647, SDGF
Chicken
Antigen affinity chromatography
RUO
374
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Western Blot
Unconjugated
P15514
AREG
Synthetic peptide corresponding to human Amphiregulin.
50 μg
Primary
Human, Non-human Primate
Antibody
IgY
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit