Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF-3 Mouse, Unlabeled, Clone: 41, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610784

Catalog No. BDB610784


Add to cart

Description

Description

The ADP-ribosylation factors (ARFs) belong to the multigene family of small GTPases capable of activating cholera toxin. ARFs fall into three different classes: Class I is composed of ARF-1, ARF-2, and ARF-3; Class II consists of ARF-4 and ARF-5; and Class III includes ARF6. Unique to ARFs is their lack of intrinsic GTP hydrolysis activity, a high affinity for GDP in a Mg2+-dependent manner, and phospholipid requirement for nucleotide exchange. ARFs are involved in intravesicular acidification of microsomal vesicles, endosome fusion, nuclear membrane assembly, and formation of clathrin-coated vesicles. In addition, GTP and ARF-3 are required for the activation of phospholipase D (PLD), an early cellular response triggered by the binding of receptors on the cell surface in response to numerous extracellular signals. Although predominantly cytosolic, ARF-3 can be translocated to cellular membranes upon cellular stimulation. The transition between the GDP-bound ARF-3 to the GTP-ARF-3 is facilitated by a high molecular weight guanine nucleotide-exchange factor sensitive to brefeldin. ARF-3 has been reported to be the most abundant in brain, kidney, and liver.

Specifications

Specifications

ARF-3
41
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
ADP Ribosylation Factor-3
Affinity Purified
Human ARF3 aa. 1-181
50μg
Cell Biology
Primary
Canine, Chicken, Frog, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit