Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522271

Catalog No. PIPA522271

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA5-22271 targets ARF1 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. The PA5-22271 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 162 (P84077) of ARF1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

ARF1
Polyclonal
Unconjugated
ARF1
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11840, 375
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.6 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
P84077, P84078
ARF1
Recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 162 (P84077) of ARF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit