Learn More
ARF1 Rabbit anti-Canine, Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA1127
Catalog No. PIPA1127
Add to cart
Description
Description
PA1-127 has been successfully used in Western blot in human, mouse, and canine samples, and in immunofluorescence and immunoprecipitation applications in human samples.
ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
Arf1
Polyclonal
Unconjugated
ARF1
Liquid
ARF1
Synthetic peptide DWLSNQLRNQK
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
P84077, P84078
ADP-ribosylation factor 1
Rabbit
IgG
100 μg
-20°C
Primary
100856517, 11840, 375