Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521961

Catalog No. PIPA521961


Add to cart

Description

Description

PA5-21961 targets ARF7 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-21961 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 192 of ARL14 (Uniprot IDNo.Q8N4G2). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

ARF7
Polyclonal
Unconjugated
ARL14
Liquid
ARL14
Recombit fragment corresponding to a region within amino acids 1 and 192 of ARL14 (Uniprot IDNo.Q8N4G2)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8N4G2
ARF7, ADP-ribosylation factor 7, ADP-ribosylation factor-like protein 14
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
80117
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit