Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARFGAP3, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131452

Catalog No. PIPA131452

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a GTPase-activating protein (GAP) which associates with the Golgi apparatus and which interacts with ADP-ribosylation factor 1 (ARF1). The encoded protein promotes hydrolysis of ARF1-bound GTP and is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles. Dissociation of the coat proteins is required for the fusion of these vesicles with target compartments. The activity of this protein is stimulated by phosphoinosides and inhibited by phosphatidylcholine. Two transcript variants encoding different isoforms have been found for this gene.

PA1-31452 has been successfully used in Western blot applications. Detects a band around 55 - 60kDa in mouse brain samples.The PA1-31452 immunogen is a peptide with sequence NGVVTSIQDRYGS, from C-Terminus of the protein sequence according to NP_055385.
Specifications

Specifications

ARFGAP3
Polyclonal
Unconjugated
ARFGAP3
Purified
ARFGAP3
Peptide with sequence NGVVTSIQDRYGS, from C-Terminus of the protein sequence according to NP_055385
IgG fraction
RUO
Antibody
Polyclonal
Human
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q9NP61
RP11-261N11.1, ARF1GAP, HRIHFB2281, MGC39924
Goat
IgG
50μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
26286
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit