Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arg3.1 Sheep anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen OSA00014G

Catalog No. PIOSA00014G


Add to cart

Description

Description

Reconstitute with 500 µL of distilled water. Centrifuge to remove any insoluble material.

Required for consolidation of synaptic plasticity as well as formation of long-term memory. Regulates endocytosis of AMPA receptors in response to synaptic activity. Required for homeostatic synaptic scaling of AMPA receptors.
Specifications

Specifications

Arg3.1
Polyclonal
PBS with no preservative
Q7LC44, Q9WV31, Q63053
Activity-regulated cytoskeleton-associated protein, Activity-regulated gene 3.1 protein homolog, KIAA0278, Arg3.1/ARC,
Sheep
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11838, 23237, 54323
Immunohistochemistry, Western Blot
Unconjugated
ARC
Lyophilized
ARC
A synthetic peptide from human Arg3.1
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit