Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arginase I Mouse, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610709

 View more versions of this product

Catalog No. BDB610709


Add to cart

Description

Description

Arginase converts arginine into urea plus ornithine, the final step in urea synthesis. Two different isoforms (I&II) have been isolated with approximately 60% homology at the nucleotide level. While type II is present in many tissues, Arginase I is expressed exclusively in liver. In cultured macrophages, as well as in vivo, Arginase I is induced with nitric oxide synthase (NOS) and the arginase I transactivator C/EBPβ in response to lipopolysaccharide. This response occurs in a dose and time-dependent manner. While the mRNA for NOS appears as early as 2h after treatment, mRNA levels for arginase I peak after twelve hours of lipopolysaccharide treatment. Since the synthesis of nitric oxide by NOS requires arginine, the delayed induction of arginase I may be necessary for the regulation of NOS activity.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Arginase I
19
Unconjugated
Affinity Purified
Human Arginase I aa. 53-207
150μg
Cell Biology
Primary
Drosophila, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.