Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARGLU1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526298

Catalog No. PIPA526298

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

ARGLU1 belongs to the UPF0430 family. There are two named isoforms.
Specifications

Specifications

ARGLU1
Polyclonal
PBS with 0.09% sodium azide
Q9NWB6
arginine and glutamate-rich protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55082
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ARGLU1
Liquid
ARGLU1
KLH conjugated synthetic peptide between 48-74 amino acids from the N-terminal region of human ARGLU1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit