Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHE Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514842

Catalog No. PIPA514842


Add to cart

Description

Description

This antibody is predicted to react with porcine and rat based on sequence homology.

Members of the Rho family of Ras-related GTPases, such as ARHE, regulate the organization of the actin cytoskeleton in response to extracellular growth factors. Like Ras (MIM 190020), Rho family members appear to cycle between an inactive GDP-bound form and an active GTP-bound form. Three major regulators of Rho activity have been identified: RhoGDIs, which interact with the GDP-bound Rho proteins to keep them in a resting complex (see MIM 601925); GEFs, which promote GDP/GTP exchange leading to activation of Rho proteins (see MIM 601855); and GAPs, which stimulate GTP hydrolysis and return the activated Rho protein to its inactive form (see MIM 602680).
Specifications

Specifications

ARHE
Polyclonal
PBS with 0.09% sodium azide
P61587, P61588
Rho-related GTP-binding protein RhoE [Precursor]; Rho family GTPase 3; Rnd3; Rho8; MemB protein; RND3; ARHE; RHO8; RHOE
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
390, 74194
Immunohistochemistry, Western Blot
Unconjugated
RND3
Liquid
RND3
KLH conjugated synthetic peptide between 133-165 amino acids from the central region of human ARHE
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit