Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP17 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524250

Catalog No. PIPA524250

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA (MIM 165390), RAC1 (MIM 602048), and CDC42 (MIM 116952).
Specifications

Specifications

ARHGAP17
Polyclonal
PBS with 0.09% sodium azide
Q68EM7
MST066, MST110, MSTP038, MSTP066, MSTP110, NADRIN, PP367, PP4534, RICH1, RICH1B, WBP15, RICH-1, RhoGAP interacting with CIP4 homologs 1, neuron-associated developmentally regulated protein, rho GTPase-activating protein 17, rho-type GTPase-activating protein 17, rhoGAP interacting with CIP4 homologs protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55114
Flow Cytometry, Western Blot
Unconjugated
ARHGAP17
Liquid
ARHGAP17
KLH conjugated synthetic peptide between 41-70 amino acids from the N-terminal region of human ARHGAP17
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit