Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP25 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524791

Catalog No. PIPA524791

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ARHGAP25 encode negative regulators of Rho GTPases (see ARHA; MIM 165390), which are implicated in actin remodeling, cell polarity, and cell migration.
Specifications

Specifications

ARHGAP25
Polyclonal
PBS with 0.09% sodium azide
P42331, Q8BYW1
KAIA0053, rho GTPase-activating protein 25, rho-type GTPase-activating protein 25
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
232201, 9938
Immunohistochemistry, Western Blot
Unconjugated
ARHGAP25
Liquid
ARHGAP25
KLH conjugated synthetic peptide between 193-219 amino acids from the central region of human ARHGAP25
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit