Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP30 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524919

Catalog No. PIPA524919

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ARHGAP30 is GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications

Specifications

ARHGAP30
Polyclonal
PBS with 0.09% sodium azide
Q7Z6I6, Q640N3
rho GTPase-activating protein 30, rho-type GTPase-activating protein 30
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
226652, 257106
Flow Cytometry, Western Blot
Unconjugated
ARHGAP30
Liquid
ARHGAP30
KLH conjugated synthetic peptide between 784-812 amino acids from the C-terminal region of human ARHGAP30
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit