Learn More
ARHGDIA, Rabbit, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of ARHGDIA.
Manufacturer: Abnova Corporation PAB18433
Description
Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA (Leffers et al., 1993 [PubMed 8262133]).[supplied by OMIM
Specifications
ARHGDIA | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of ARHGDIA. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
P52565 | |
GDIA1|MGC117248|RHOGDI|RHOGDI-1 | |
Rabbit | |
Affinity purification | |
RUO | |
Primary | |
396 | |
Human, Murine, Rat |
ELISA, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
ARHGDIA | |
Liquid | |
ARHGDIA | |
A synthetic peptide corresponding to human ARHGDIA. | |
100 ug | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
This antibody is specific to ARHGDIA. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only