Learn More
anti-ARHGEF5, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP215457
Catalog No. NBP215457
Add to cart
Description
Description
ARHGEF5 Polyclonal specifically detects ARHGEF5 in Human samples. It is validated for Western Blot.
Specifications
Specifications
ARHGEF5
0.54mg/mL
Western Blot 1:500-1:3000
Q12774
ephexin-3, GEF5, Guanine nucleotide regulatory protein TIM, Oncogene TIM, P60, p60 TIM, Rho guanine nucleotide exchange factor (GEF) 5, rho guanine nucleotide exchange factor 5, TIM1, TIMDKFZp686N1969, Transforming immortalized mammary oncogene
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
ARHGEF5
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human GEF5. The exact sequence is proprietary.
177 kDa
0.1 ml
Cancer, GPCR
Primary
7984