Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGEF9 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512281

Catalog No. PIPA512281

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and mouse based on sequence homology.

ARHGEF9 belongs to a family of Rho-like GTPases that act as molecular switches by cycling from the active GTP-bound state to the inactive GDP-bound state. These proteins are key regulators of the actin cytoskeleton and are involved in cell signaling.
Specifications

Specifications

Arhgef9
Polyclonal
PBS with 0.09% sodium azide
O43307, Q9QX73
Rho guanine nucleotide exchange factor 9; Rac/Cdc42 guanine nucleotide exchange factor 9; Collybistin; Arhgef9
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23229, 66013
Immunohistochemistry, Western Blot
Unconjugated
ARHGEF9
Liquid
ARHGEF9
KLH conjugated synthetic peptide between 274-303 amino acids from the central region of human Arhgef9
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit