Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ARID2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126614

Catalog No. NBP126614


Add to cart

Description

Description

ARID2 Polyclonal antibody specifically detects ARID2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

ARID2
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 5-10 ug/mg lysate
NP_689854.2
ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee and Crab-eating macaque.
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
ARID2
The immunogen recognized by this antibody maps to a region between residue 1600 and 1650 of human AT rich interactive domain 2 using the numbering given in entry NP_689854.2 (GeneID 196528).
Immunogen affinity purified
RUO
Primary
196528
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only