Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AT rich interactive domain 3A (BRIGHT-like), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ARID3A protein.

Manufacturer:  Abnova Corporation H00001820B01

Catalog No. 89-005-262


Add to cart

Description

Description

This gene encodes a member of the ARID (AT-rich interaction domain) family of DNA binding proteins. It was found by homology to the Drosophila dead ringer gene, which is important for normal embryogenesis. Other ARID family members have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation, and possibly in chromatin structure modification. [provided by RefSeq

Sequence: MKLQAVMETLLQRQQRARQELEARQQLPPDPPAAPHGRARAAPDEDREPESARMQRAQMAALAAMRAAAAGLGHPASPGGSEDGPPGSEEEDAAREGTPGSPGRGREGPGEEHFEDMASDEDMKPKWEEEEMEEDLGEDEEEEEEDYEDEEEEEDEEGLGPPGPASLGTTALFPRKAQPPQAFRGDGVPRVLGGQERPGPGPAHPGGAAHVAPQLQPPDHGDWTYEEQFKQLYELDGDPKRKEFLDDLFSFMQKRGTPVNRIPIMAKQVLDLFMLYVLVTEKGGLVEVINKKLWREITKGLNLPTSITSAAFTLRTQYMEYLYPYECEKRGLSNPNELQAAIDSNRREGRRQSFGGSLFAYSPGGAHGMLSSPKLPVSSLGLAASTNGSSITPAPKIKKEEDSAIPITVPGRLPVSLAGHPVVAAQAAAVQAAAAQAAVAAQAAALEQLREKLESAEPPEKKMALVADEQQRLMQRALQQNFLAMAAQLPMSIRINSQASESRQDSAVNLTGTNGSNSISMSVEINGIMYTGVLFAQPPAPTPTSAPNKGGGGGGSSSSNAGGRGGNTGTSGGQAGPAGLSTPSTSTSNNSLP
Specifications

Specifications

AT rich interactive domain 3A (BRIGHT-like)
Unconjugated
No additive
BC060828.1
BRIGHT|DRIL1|DRIL3|E2FBP1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1820
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ARID3A protein.
ARID3A
Serum
ARID3A
ARID3A (AAH60828.1, 1 a.a. ~ 593 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit