Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ARK5/NUAK1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136937

Catalog No. NBP136937


Add to cart

Description

Description

ARK5/NUAK1 Polyclonal antibody specifically detects ARK5/NUAK1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications

Specifications

ARK5/NUAK1
0.5 mg/mL
Flow Cytometry 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Peptide ELISA Detection limit 1:8000
NP_055655.1
AMPK-related protein kinase 5, ARK5AMP-activated protein kinase family member 5, EC 2.7.11, EC 2.7.11.1, KIAA0537NUAK family SNF1-like kinase 1, NUAK family, SNF1-like kinase, 1, NuaK1
Goat
IgG
0.1 mg
Protein Kinase
Primary
9891
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
NUAK1
Peptide with sequence C-QNRPRPQYLKRYRNR corresponding to internal region according to NP_055655.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Human. Expected from sequence similarity: Rat.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only