Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL8A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523671

Catalog No. PIPA523671


Add to cart

Specifications

Specifications

ARL8A
Polyclonal
PBS with 0.09% sodium azide
Q96BM9, Q8VEH3
ARL10B, GIE2, ADP-ribosylation factor-like 10B, ADP-ribosylation factor-like protein 8A, novel small G protein indispensable for equal chromosome segregation 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
127829, 68724
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ARL8A
Liquid
ARL8A
KLH conjugated synthetic peptide between 116-142 amino acids from the central region of human ARL8A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit