Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-AMICA/JAML, APC, Clone: 4E10, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143309APC

Catalog No. NBP143309J


Add to cart

Description

Description

AMICA/JAML Monoclonal antibody specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

AMICA/JAML
40000000000
Flow Cytometry
Affinity Purified
AMICA1
The immunogen for this antibody was JAML.
Protein A or G purified
RUO
Primary
120425
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
Armenian Hamster
IgG
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only