Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD27/TNFRSF7, PE, Clone: LG.7F9, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143428PE

Catalog No. NBP143428PE


Add to cart

Description

Description

CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Product Image.
Specifications

Specifications

CD27/TNFRSF7
LG.7F9
Flow Cytometry
Affinity Purified
CD27
The immunogen for this antibody was CD27.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Armenian Hamster
IgG
0.1 ml
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity
Primary
939
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only