Learn More
anti-CD28, FITC, Clone: PV-1, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127933
Catalog No. NBP127933
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD28 Monoclonal antibody specifically detects CD28 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.
Specifications
Specifications
CD28
PV-1
FITC
PBS with 0.05% Sodium Azide
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Armenian Hamster
IgG2
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Murine CD28
Flow Cytometry, Immunoprecipitation, In vitro assay
0.5mg/mL
Flow Cytometry <= 3 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay
Purified
CD28
C57BL/6N mouse T cell lymphoma EL4 cell line
Protein A or G purified
RUO
Primary
940
Mouse