Learn More
anti-CD28, PE, Clone: PV-1, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127935
Catalog No. NBP127935
Add to cart
Description
Description
CD28 Monoclonal antibody specifically detects CD28 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.
Specifications
Specifications
CD28
PV-1
PE
PBS and a stabilizing agent with Sodium Azide
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Armenian Hamster
IgG
0.2 mg
Store at 4C. Do not freeze.
Monoclonal
Murine CD28
Flow Cytometry, Immunoprecipitation, In vitro assay
0.1 mg/mL
Flow Cytometry <= 0.3 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay
Purified
CD28
C57BL/6N mouse T cell lymphoma EL4 cell line
Protein A or G purified
RUO
Primary
940
Mouse