Learn More
anti-CD30/TNFRSF8, Clone: 2SH12-5F, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP127937
Catalog No. NBP127937
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD30/TNFRSF8
2SH12-5F
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Armenian Hamster
IgG Kappa
0.5 mg
Embryonic Stem Cell Markers, Stem Cell Markers
Primary
943
Mouse
Flow Cytometry, In vitro assay, Blocking Assay
0.5 mg/mL
Flow Cytometry 3 ug/10^6 cells, In vitro assay, Blocking/Neutralizing 50-200 molar excess
Purified
TNFRSF8
Mouse CD30-mouse IgG1 fusion protein
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Murine CD30, Mr 52 kDa (core peptide)