Learn More
anti-CD40 Ligand/TNFSF5, FITC, Clone: MR1, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127967
Catalog No. NBP127967
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD40 Ligand/TNFSF5 Monoclonal antibody specifically detects CD40 Ligand/TNFSF5 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay.
Specifications
Specifications
CD40 Ligand/TNFSF5
MR1
FITC
PBS with 0.05% Sodium Azide
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Armenian Hamster
IgG1 Kappa
0.25 mg
Store at 4C in the dark.
Monoclonal
CD154 (Mouse CD40 ligand/gp39), Mr 39-kDa
Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay
0.5mg/mL
Western Blot, Flow Cytometry <= 2 ug/10^6 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Purified
CD40LG
Activated mouse Th1 clone D1.6
Protein A or G purified
RUO
Primary
959
Mouse