Learn More
anti-CD69, Alexa Fluor 700, Clone: H1.2F3, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128011AF700
Catalog No. NBP128011Z
Add to cart
Description
Description
CD69 Monoclonal antibody specifically detects CD69 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD69
H1.2F3
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CD69
Mouse dendritic epidermal cell line Y245
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse CD69/Very Early Activation Antigen (VEA), Mr 85 kDa (unreduced)
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
Armenian Hamster
IgG1
0.25 ml
Innate Immunity
Primary
969
Mouse