Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD69, PE, Clone: H1.2F3, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128011PE

Catalog No. NBP128011PE


Add to cart

Description

Description

CD69 Monoclonal antibody specifically detects CD69 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD69
H1.2F3
Flow Cytometry
Purified
CD69
Mouse dendritic epidermal cell line Y245
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse CD69/Very Early Activation Antigen (VEA), Mr 85 kDa (unreduced)
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
Armenian Hamster
IgG1
0.25 ml
Innate Immunity
Primary
969
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only