Learn More
anti-CD79B, Clone: HM79-12, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128140
Catalog No. NBP128140
Add to cart
Description
Description
CD79B Monoclonal antibody specifically detects CD79B in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD79B
HM79-12
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Armenian Hamster
IgG Kappa
0.5 mg
Store at 4C. Do not freeze.
Monoclonal
beta chain of the mouse B-cell receptor, Mr 23 kDa
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
0.5 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Purified
CD79B
CD79alpha/CD79B heterodimers purified from mouse (BALB/c x NZB)F1 B cell lymphoma cells, WEHI231
Protein A or G purified
RUO
Primary
974
Mouse