Learn More
anti-CD81, FITC, Clone: 2F7, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128137
Catalog No. NBP128137
Add to cart
Description
Description
CD81 Monoclonal antibody specifically detects CD81 in Human, Mouse, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Immunocytochemistry.
Specifications
Specifications
CD81
2F7
FITC
PBS with 0.05% Sodium Azide
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Armenian Hamster
IgG3 Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse CD81 (Mr 25 kDa), a member of the transmembrane 4 integral membrane protein family
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, In vitro assay
0.5mg/mL
Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Immunocytochemistry
Purified
CD81
Mouse epithelial cell line PAM212
Protein A or G purified
RUO
Primary
975
Hamster, Human, Mouse