Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-Fas Ligand/TNFSF6, FITC, Clone: MFL4, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP128810
Catalog No. NBP128810
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Fas Ligand/TNFSF6
MFL4
FITC
Affinity Purified
The immunogen for this antibody was CD95 Ligand.
0.1mg
Primary
356
Flow Cytometry
0.1mg/mL
PBS, pH 7.4 with 1% BSA
Armenian Hamster
IgG
RUO
Monoclonal
Murine, Rat