Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Fc epsilon RI, Alexa Fluor 647, Clone: MAR-1, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143279AF647

Catalog No. NBP143279Y


Add to cart

Description

Description

Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

Fc epsilon RI
43160
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Affinity Purified
FCER1A
The immunogen for this antibody was FceRI alpha.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide
Armenian Hamster
IgG
0.1 ml
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primary
2205
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only