Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc epsilon RI Antibody (43160), DyLight 680, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP143279FR
Description
Fc epsilon RI Monoclonal specifically detects Fc epsilon RI in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Fc epsilon RI | |
Monoclonal | |
DyLight 680 | |
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
Armenian Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
IgG |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
43160 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
FCER1A | |
The immunogen for this antibody was FceRI alpha. | |
0.1 mL | |
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
2205 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction