Learn More
anti-Integrin alpha E/CD103, PE, Clone: 2E7, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128126
Catalog No. NBP128126
Add to cart
Description
Description
Integrin alpha E/CD103 Monoclonal antibody specifically detects Integrin alpha E/CD103 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay.
Specifications
Specifications
Integrin alpha E/CD103
20000000
PE
PBS and a stabilizing agent with Sodium Azide
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Armenian Hamster
IgG2a
0.1 mg
Innate Immunity
Primary
3682
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, In vitro assay
0.1 mg/mL
Flow Cytometry 0.2 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Purified
ITGAE
C57BL/6J mouse intestinal intraepithelial lymphocytes (IEL)
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The CD103/alpha IELbeta7 integrin complex (Mr 160 and 115 kDa peptides).