Learn More
anti-TCR alpha/beta, Alexa Fluor 405, Clone: H57-597, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP130415AF405
Catalog No. NBP130415W
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
TCR alpha/beta
H57-597
50mM Sodium Borate
Armenian Hamster
IgG
RUO
Monoclonal
The hamster monoclonal antibody H57-597 reacts specifically with beta subunit of TCR alpha/beta. This antibody does not crossreact with TCR gamma/delta type and has been used as TCR alpha/beta phenotypic marker.
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Alexa Fluor 405
Affinity Purified
Affinity purified TCR from DO-11.10 T cell hybridoma
100 Tests
Primary
6955
Murine