Learn More
anti-TCR beta, Biotin, Clone: H57-597, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128062
Catalog No. NBP128062
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Immunocytochemistry.
Specifications
Specifications
TCR beta
H57-597
Biotin
PBS with 0.05% Sodium Azide
T cell receptor beta chain, TCRB, TCRbeta, Tib
Armenian Hamster
IgG2
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
The Armenian hamster monoclonal antibody H57-597 reacts specifically with beta subunit of TCR alpha/beta. This antibody does not crossreact with TCR gamma/delta type and has been used as TCR alpha/beta phenotypic marker.
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
0.5mg/mL
Flow Cytometry <= 2 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Immunocytochemistry
Purified
Tcrb
Affinity purified TCR from DO-11.10 T cell hybridoma
IgG purified
RUO
Primary
21577
Mouse