Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TNF RII/TNFRSF1B, PerCP, Clone: RM0012-6F13, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NB11085470PCP

Catalog No. NB11085470P



Description

Description

TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

TNF RII/TNFRSF1B
RM0012-6F13
Flow Cytometry
Purified
TNFRSF1B
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from an Armenian Hamster immunized with mouse Recombinant extracellular domain of mouse TNFR-II protein.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This is specific to the extracellular domain of mouse TNFR-II protein.
Flow Cytometry, ELISA, Immunoprecipitation
PerCP
PBS with 0.05% Sodium Azide
CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Armenian Hamster
IgG
0.1 ml
Apoptosis, Cancer, Cytokine Research, Zinc Finger
Primary
7133
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only